Skip to Content
Merck
All Photos(1)

Documents

PX0397

Supelco

Perchloric acid

OmniTrace

Synonym(s):

aa, PCA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HClO4
CAS Number:
Molecular Weight:
100.46
MDL number:
UNSPSC Code:
12352301

Quality Level

product line

OmniTrace®

bp

198.7 °C/1013 hPa

mp

-18 °C

shipped in

ambient

storage temp.

room temp

SMILES string

OCl(=O)(=O)=O

InChI

1S/ClHO4/c2-1(3,4)5/h(H,2,3,4,5)

InChI key

VLTRZXGMWDSKGL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

OmniTrace
PX0397-1 and PX0397-2 are packaged in poly coated glass bottles

Legal Information

OMNITRACE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 1 - Skin Corr. 1A - STOT RE 2

Target Organs

Thyroid

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service