Skip to Content
Merck
All Photos(2)

Key Documents

MABS1268

Sigma-Aldrich

Anti-MCRS1 Antibody, clone DON193B/C9

clone DON193B/C9, from rat

Synonym(s):

Microspherule protein 1, 58 kDa microspherule protein, Cell cycle-regulated factor p78, INO80 complex subunit J, MCRS2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

DON193B/C9, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... MCRS1(10445)

General description

Microspherule protein 1 (UniProt: Q96EZ8; also known as 58 kDa microspherule protein, Cell cycle-regulated factor p78, INO80 complex subunit J, MCRS1) is encoded by the MCRS1 (also known as INO80Q, MSP58) gene (Gene ID: 10445) in human. MCRS1 is a nucleolar protein that is detected in testis, and at lower levels in spleen, thymus, prostate, uterus, small intestine, colon and leukocytes. Its highest levels are reported in the early S phase and it becomes undetectable in the G2 phase. Four isoforms of MCRS1 have been reported that are produced by alternative splicing. MCRS1 contains a nucleolar localization motif (aa 113-123), a coiled-coil domain (aa 301-335), and a forkhead associated (FHA) domain (aa 363-419). MCRS1 exhibits transforming activity, which is suppressed by phosphatase and tensin homologue (PTEN). It can modulate the transcription repressor activity of DAXX by recruiting it to the nucleolus. MCRS1 forms a complex with Mi-2 beta, and RET finger protein (RFP) and co-localize in the Nucleolus. This complex activates rRNA promoter activity. (Ref.: Shimono, K., et al. (2005). J. Biol. Chem. 280(47); 39436-39447).

Specificity

Clone DON193B/C9 detects Microspherule protein 1 in human cells.

Immunogen

A mixture of GST-tagged full length recombinant human Microspherule protein 1 and GST-tagged C- and N-terminal recombinant fragments of human Microspherule protein 1.
Epitope: unknown

Application

Anti-MCRS1, clone DON193B/C9, Cat. No. MABS1268, is a rat monoclonal antibody that detects Microspherule protein 1 and has been tested for use in Western Blotting.
Research Category
Signaling
Western Blotting Analysis: A representative lot detected MCRS1 in human tonsil, human spleen, HeLa cell, and Ramos cell lysates (Courtesy of Giovanna Roncador, Ph.D., CNIO, Madrid, Spain).

Quality

Evaluated by Western Blotting in HCT116 cell lysate.

Western Blotting Analysis: 4 µg/mL of this antibody detected MCRS1 in 10 µg of HCT116 cell lysate.

Target description

~52 kDa observed; 51.80 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service