Skip to Content
Merck
All Photos(2)

Documents

MABN879

Sigma-Aldrich

Anti-phospho-Amyloid beta Antibody (Ser26), clone 5H11C10

clone 5H11C10, from mouse

Synonym(s):

Aβ peptide, Ser26 phosphorylated, Abeta peptide, Ser26 phosphorylated, Amyloid β peptide, Ser26 phosphorylated, Aβ1-40, Ser26 phosphorylated, Aβ1-42, Ser26 phosphorylated, Abeta1-40, Ser26 phosphorylated, Abeta1-42, Ser26 phosphorylat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5H11C10, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

phosphorylation (pSer26)

Gene Information

human ... APP(351)

General description

Amyloid beta A4 protein (UniProt P05067; also known as ABPP, Alzheimer disease amyloid protein, Amyloid precursor protein, APP, APPI, Cerebral vascular amyloid peptide, CVAP, PN-II, PreA4, Protease nexin-II) is encoded by the APP (also known as A4, AD1) gene (Gene ID 351) in human. Amyloid precursor protein (APP) is initially produced with a signal peptide sequence (a.a. 1-17), the removal of which yields the mature protein with a large extracellular portion (a.a. 18-699), followed by a transmembrane segment (a.a. 700-723) and a cytoplasmic (a.a. 724-770) tail. APP can be further processed by the α-, β-, and γ-secretases in two alternative processing pathways. In the non-amyloidogenic pathway, APP is first cleaved by the plasma membrane-localized α-secretase to generate an N-terminal extracellular sAPPα fragment (a.a. 18-687) and a membrane-bound C-terminal fragment C83 (CTFα), which can be further cleaved by γ-secretase to produce a non-toxic small peptide p3 and a cytoplasmic APP intracellular domain (AICD). In the amyloidogenic pathway, APP undergoes β-cleavage in BACE-1 (β-site APP-cleaving enzyme)-enriched endosomes to generate an N-terminal extracellular sAPPβ fragment (a.a. 18-671) and a membrane-bound C-terminal fragment C99 (CTFβ). Subsequent cleavage of C99 by γ-secretase releases the amyloid β peptides, Aβ1-42 (672-713) & Aβ1-40 (672-711), and AICD. Aβ accumulation in the cortical and hippocampal regions of the brain is a major pathological feature of Alzheimer′s disease (AD). Aβ serine phosphorylation is shown to promote Aβ aggregation into oligomeric and fibrillar assemblies and to prevent Aβ proteolytic clearance by certain proteases.

Specificity

Clone 5H11C10 recognizes Aβ1-40 and Aβ1-42 peptides with phosphorylated Ser26, but not the non-phosphorylated peptides.

Immunogen

KLH-conjugated linear peptide corresponding to human amyloid beta peptide target region sequence with phosphorylated Ser26.

Application

Anti-phospho-Amyloid beta (Ser26), clone 5H11C10, Cat. No. MABN879, is a highly specific mouse monoclonal antibody, that targets amyloid beta peptides Ser26 phosphorylation and has been tested in ELISA, Immunofluorescence, Western Blotting.
Western Blotting Analysis: 2 µg/mL from a representative lot detected monomeric and dimeric forms of synthetic A 1-40 peptide with phosphorylated Ser26, but not the non-phosphorylated A 1-40 peptide (Courtesy of Dr. Kumar Sathish, University of Bonn, Germany)

Western Blotting Analysis: 2 µg/mL from a representative lot detected Ser26-phosphorylated oligomeric amyloid beta (pA ) peptides in brain extracts from 8-month old APP/PS1KI transgenic mice, but not in extracts from age-matched non-transgenic mice (Courtesy of Dr. Kumar Sathish, University of Bonn, Germany).

Note: Formic acid (88%) treatment following heat retrieval is recommended for immunohistochemical detection of aggregated intraneuronal Abeta peptides in brain sections (Kumar, S., et al. (2013). Acta Neuropathol. 125(5):699-709; Christensen, D.Z., et al. (2009). Brain Res. 1301:116-125).

Quality

Identity Confirmation by Isotyping Test.

Isotyping Analysis: The identity of this monoclonal antibody is confirmed by isotyping test to be IgG1κ.

Target description

~4/8 (monomer/dimer) and >188 (oligomer) kDa observed. 4.514/9.028 kDa (Abeta1-42 monomer/dimer), 4.330/8.660 kDa (Abeta1-40 monomer/dimer) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service