Recommended Products
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
Unknown, polyclonal
mol wt
calculated mol wt 38.92 kDa
observed mol wt ~37 kDa
purified by
affinity chromatography
species reactivity
mouse
species reactivity (predicted by homology)
rat
packaging
antibody small pack of 100 μg
technique(s)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable
isotype
IgG
epitope sequence
C-terminal half
Protein ID accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
rabbit ... Pun1sw(12057)
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in NRL knockout mouse retina lysate
Western Blotting Analysis: A 1:10,000 dilution of this antibody detected S-Opsin in NRL knockout mouse retina tissue lysate.
Tested Applications
Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution from a representative lot detected S-Opsin in mouse retina tissue sections.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected S-Opsin in Mouse Eye tissue lysate.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service