Skip to Content
Merck
All Photos(1)

Documents

8.20872

Sigma-Aldrich

di-Sodium DL-malate

98-101% (perchloric acid titration)

Synonym(s):

di-Sodium DL-malate, DL-Malic acid disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOOCCH(OH)CH2COONa
CAS Number:
Molecular Weight:
178.05
MDL number:
UNSPSC Code:
12352107
EC Index Number:
211-627-6
NACRES:
NA.22

Quality Level

Assay

98-101% (perchloric acid titration)

form

powder

storage temp.

2-30°C

InChI

1S/C4H6O5.2Na/c5-2(4(8)9)1-3(6)7;;/h2,5H,1H2,(H,6,7)(H,8,9);;/q;2*+1/p-2

InChI key

WPUMTJGUQUYPIV-UHFFFAOYSA-L

General description

for synthesis

Analysis Note

Assay (perchloric acid titration): 98 - 101 %
Identity (NMR-spectrum): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service