Skip to Content
Merck
All Photos(1)

Documents

81129-M

SAFC

EX-CYTE® Growth Enhancement Media Supplement

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
eCl@ss:
32011401

form

liquid

Quality Level

shelf life

24 mo.

manufacturer/tradename

Ex-cyte®

General description

Product Source: Bovine; Adult Bovine Serum

Storage and Stability

refrigerated

Legal Information

EX-CYTE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For further manufacturing use only. Not for human or animal consumption.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

A custom-designed cost model is used to explore the economics of vaccine manufacturing across several different modalities including mRNA. The model enables greater process understanding, simulates bottlenecks, and helps to optimize production efficiency.

Influenza vaccines are commonly made using egg-based and cell-based manufacturing strategies. Find step-by-step information on the manufacturing process for each method.

Learn more one the attenuated viral vaccines manufacturing process: cell culture, clarification, nuclease treatment, chromatography, and sterile filtration.

This technical article breaks down the steps of upstream and downstream bioprocessing and formulation of virus-like particle vaccines.

Related Content

This technical article breaks down the adenovirus vaccine manufacturing process and provides a case study on developing an accelerated and cost-effective single-use adenoviral vector vaccine.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service