Skip to Content
Merck
All Photos(1)

Documents

662047

Sigma-Aldrich

Anti-Uncoupling Protein-2 (UCP-2) (144-157) Rabbit pAb

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

0.1% sodium azide as preservative

species reactivity

human, mouse, rat

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−70°C

target post-translational modification

unmodified

Gene Information

human ... UCP2(7351)

General description

Protein A purified rabbit polyclonal antibody. Recognizes the UCP-2 protein.
Recognizes the UCP-2 protein.

Immunogen

a synthetic peptide [(C)QARAGGGRRYQSTV] corresponding to amino acids 144-157 of human and mouse UCP-2

Application

ELISA (1:100,000)

Immunoblotting (1:1000)

Immunocytochemistry (1:500)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Other Notes

Does not react with UCP-1 or UCP-3. Since this antigen sequence is highly conserved, cross-reactivity across a broad range of species is expected. Variables associated with assay conditions will dictate the proper working dilution.
Stuart, J.A., et al. 2001. J. Biol. Chem.276, 18633.
Gimeno, R.E., et al. 1997. Diabetes46, 900.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jay D Turner et al.
Circulation research, 106(4), 730-738 (2010-01-09)
Uncoupling protein (UCP)2 is a mitochondrial inner membrane protein that is expressed in mammalian myocardium under normal conditions and upregulated in pathological states such as heart failure. UCP2 is thought to protect cardiomyocytes against oxidative stress by dissipating the mitochondrial
P Brauner et al.
Pediatric research, 49(3), 440-447 (2001-03-03)
Hepatic hematopoiesis is prominent during fetal life and ceases around birth. In rodent liver, the decline of the hepatic hematopoiesis starts abruptly at birth being accompanied by a decrease of mitochondrial uncoupling protein 2 (UCP2) expression in monocytes/macrophages, whereas hepatocytes
C Y Zhang et al.
Cell, 105(6), 745-755 (2001-07-07)
beta cells sense glucose through its metabolism and the resulting increase in ATP, which subsequently stimulates insulin secretion. Uncoupling protein-2 (UCP2) mediates mitochondrial proton leak, decreasing ATP production. In the present study, we assessed UCP2's role in regulating insulin secretion.
Jan Ježek et al.
Antioxidants & redox signaling, 23(12), 958-972 (2015-05-01)
Pancreatic β-cell chronic lipotoxicity evolves from acute free fatty acid (FA)-mediated oxidative stress, unprotected by antioxidant mechanisms. Since mitochondrial uncoupling protein-2 (UCP2) plays antioxidant and insulin-regulating roles in pancreatic β-cells, we tested our hypothesis, that UCP2-mediated uncoupling attenuating mitochondrial superoxide
Yi-Hsieng Samuel Wu et al.
Journal of food and drug analysis, 29(2), 375-388 (2022-06-14)
This study aims to clarify the effects of chicken liver hydrolysates (CLHs) on long-term high-fat diet (HFD)-induced insulin resistance (IR) and hepatosteatosis in mice. In vitro, the 400 μM oleic acid (OA)-added medium successfully stimulated the cellular steatosis on FL83B

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service