Skip to Content
Merck
All Photos(1)

Key Documents

454565

Sigma-Aldrich

Methyl Arachidonyl Fluorophosphonate - Calbiochem

Synonym(s):

Methyl Arachidonyl Fluorophosphonate - Calbiochem, MAFP, cPLA2α Inhibitor III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H36FO2P
Molecular Weight:
370.48
UNSPSC Code:
12352200

Assay

≥98% (TLC)

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

wet ice

storage temp.

−70°C

General description

This product has been discontinued.



A selective, active site-directed, irreversible inhibitor of both calcium-dependent and calcium-independent cytosolic (85 kDa) phospholipase A2 (PLA2) but not of secretory PLA2.

A selective, active site-directed, irreversible inhibitor of both calcium-dependent and calcium-independent cytosolic (85 kDa) phospholipase A2 (PLA2) but not secretory PLA2.

Biochem/physiol Actions

Cell permeable: no
Primary Target
calcium-dependent and calcium-independent cytosolic (85 kDa) phospholipase A2 (PLA2)
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Physical form

In methyl acetate.

Reconstitution

Prior to use, evaporate the methyl acetate and resuspend in high quality chloroform. Aliquot and immediately evaporate the chloroform. Store aliquots at -70°C. Reconstitute in DMSO just prior to use.

Other Notes

Balsinde, J., and Dennis, E.A. 1996. J. Biol. Chem. 271, 6758.
Lio, Y.C., et al. 1996. Biochim. Biophys. Acta1302, 55.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Balsinde et al.
The Journal of biological chemistry, 271(12), 6758-6765 (1996-03-22)
Receptor-stimulated arachidonic acid (AA) mobilization in P388D1 macrophages consists of a transient phase in which AA accumulates in the cell and a sustained phase in which AA accumulates in the incubation medium. We have shown previously that a secretory group
Y C Lio et al.
Biochimica et biophysica acta, 1302(1), 55-60 (1996-07-12)
Methyl arachidonyl fluorophosphonate (MAFP) has been recently reported to be a selective, active-site directed, irreversible inhibitor of the Group IV 85 kDa cytosolic phospholipase A2 (cPLA2). We have now shown that this compound also potently inhibits the Ca(2+)-independent cytosolic phospholipase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service