Skip to Content
Merck
All Photos(1)

Key Documents

344085

Sigma-Aldrich

Folimycin, Streptomyces sp.

A highly sensitive and specific inhibitor of vacuolar-type H+-ATPase (V-type; Ki = 20 pM).

Synonym(s):

Folimycin, Streptomyces sp., Concanamycin A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H75NO14
CAS Number:
Molecular Weight:
866.09
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥90% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

solubility

DMSO: soluble

shipped in

ambient

storage temp.

−20°C

InChI

1S/C46H75NO14/c1-13-16-34-28(7)37(58-38-22-33(48)43(31(10)57-38)60-45(47)53)23-46(54,61-34)30(9)41(51)29(8)42-35(55-11)18-15-17-24(3)19-26(5)39(49)32(14-2)40(50)27(6)20-25(4)21-36(56-12)44(52)59-42/h13,15-18,20-21,26-35,37-43,48-51,54H,14,19,22-23H2,1-12H3,(H2,47,53)/b16-13+,18-15+,24-17+,25-20+,36-21-/t26-,27-,28-,29+,30+,31-,32+,33-,34-,35+,37-,38+,39+,40-,41-,42-,43-,46-/m1/s1

InChI key

DJZCTUVALDDONK-HQMSUKCRSA-N

General description

A highly sensitive and specific inhibitor of vacuolar-type H+-ATPase (V-type; Ki = 20 pM). Inhibits acidification of organelles, such as lysosomes and the Golgi apparatus. Also blocks cell surface expression of viral envelope glycoproteins without affecting their synthesis. Useful for studies of intracellular protein translocation. Exhibits cytotoxic effects on a number of cell lines in a cell viability assay.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Vacuolar-type H+-ATPase
Product does not compete with ATP.
Reversible: no
Target Ki: 20 pM against vacuolar-type H+-ATPase

Warning

Toxicity: Highly Toxic (H)

Preparation Note

Further dilute with aqueous buffers just prior to use.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 year at -20°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Kane, M.D., et al. 1999. J. Neurochem.72, 1939.
Nishihara, T., et al. 1995. Biochem. Biophys. Res. Commun.212, 255.
Muroi, M., et al. 1994. Biosci. Biotech. Biochem.58, 425.
Drose, S., et al. 1993. Biochemistry32, 3902.
Muroi, M., et al. 1993. Biochem. Biophys. Res. Commun.193, 999.
Muroi, M., et al. 1993. Cell Struct. Funct.18, 139.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Eye Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Göttle et al.
Frontiers in cellular neuroscience, 15, 777542-777542 (2021-12-11)
Myelin repair in the adult central nervous system (CNS) is driven by successful differentiation of resident oligodendroglial precursor cells (OPCs) and thus constitutes a neurodegenerative process capable to compensate for functional deficits upon loss of oligodendrocytes and myelin sheaths as
Michael J Rigby et al.
Communications biology, 4(1), 454-454 (2021-04-14)
Nε-lysine acetylation in the ER lumen is a recently discovered quality control mechanism that ensures proteostasis within the secretory pathway. The acetyltransferase reaction is carried out by two type-II membrane proteins, ATase1/NAT8B and ATase2/NAT8. Prior studies have shown that reducing
Jin Rui Liang et al.
Cell, 180(6), 1160-1177 (2020-03-12)
Selective autophagy of organelles is critical for cellular differentiation, homeostasis, and organismal health. Autophagy of the ER (ER-phagy) is implicated in human neuropathy but is poorly understood beyond a few autophagosomal receptors and remodelers. By using an ER-phagy reporter and
Keiji Ibata et al.
Neuron, 102(6), 1184-1198 (2019-05-11)
Synapse formation is achieved by various synaptic organizers. Although this process is highly regulated by neuronal activity, the underlying molecular mechanisms remain largely unclear. Here we show that Cbln1, a synaptic organizer of the C1q family, is released from lysosomes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service