1.46459
Fluid Thioglycollate Medium
bottle size 250 mL , filling volume
Synonym(s):
FTM, Thioglycollate Broth
About This Item
Recommended Products
Agency
EP
JP 4.06
USP
Quality Level
Sterilization Compatibility
autoclavable compatible
form
liquid
feature
closure type crimp cap with septum
packaging
single packed of 6 × 250 mL each
bottle of
bottle capacity
250 mL
bottle size
250 mL , filling volume
pH
6.9±0.2 (25 °C)
application(s)
pharmaceutical
sterility testing
storage temp.
2-25°C
suitability
nonselective for
General description
The anaerobic atmosphere is created through reducing the redox potential of the medium by Thioglycollate and L-Cystine. Additionally, these compounds inactivate mercury and other heavy metal elements present in the environment.
The higher viscosity of the medium prevents rapid oxygen uptake and any increase in the oxygen content is indicated by the indicator, sodium resazurin which changes its color to red.
Application
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service