Skip to Content
Merck
All Photos(1)

Documents

04-932

Sigma-Aldrich

Anti-Calmodulin Binding Protein Epitope Tag Antibody, clone C16T, rabbit monoclonal

culture supernatant, clone C16T, from rabbit

Synonym(s):

calmodulin 1, calmodulin 1 (phosphorylase kinase, delta), phosphorylase kinase, delta subunit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

C16T, monoclonal

species reactivity

vertebrates

technique(s)

western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CALM1(801)

General description

Calmodulin Binding Protein (CBP) epitope tags are found in several E. coli expression vectors. Calmodulin (CaM), a small 17 kDa ubiquitous Ca2+ binding protein, acts as an intracellular Ca2+ receptor and transduces Ca2+ transients, usually in response to growth factors. It mediates the activity of a number of Ca2+ regulating enzymes, including protein kinases, phosphatases, nitric oxide synthases, and phosphodiesterase. Calmodulin is the principal intracellular calcium receptor. It is an acidic protein of 148 residues, and upon binding Ca2+, it in turn binds to short peptide sequences of many target proteins. Such interaction is thought to induce a conformational change on the target, resulting in its activation. Targets include the constitutively expressed nitric oxide synthases, the CaM kinases, and PP2B/Calcineurin.

Specificity

Recombinant proteins containing the calmodulin binding protein epitope tag.

Immunogen

Epitope: CBP epitope
KLH-conjugated, synthetic peptide corresponding to the Calmodulin Binding Protein (CBP) epitope tag found in several E. coli expression vectors.

Application

Research Category
Epitope Tags & General Use
Research Sub Category
Epitope Tags
This Anti-Calmodulin Binding Protein Epitope Tag Antibody, clone C16T is validated for use in WB for the detection of Calmodulin Binding Protein Epitope Tag.

Quality

Evaluated by Western Blot on lysates from transformed E. coli.
Western Blot Analysis: 1:2,000 dilution of this antibody detected a recombinant protein containing the calmodulin binding protein epitope tag in lysates from transformed E. coli.

Target description

The molecular weight is related to the tagged protein.

Linkage

Replaces: 05-932

Physical form

Cultured supernatant with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at -20ºC from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Samples containing the calmodulin binding protein epitope tag.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Josua Schinke et al.
PLoS genetics, 12(3), e1005949-e1005949 (2016-03-25)
DenA/DEN1 and the COP9 signalosome (CSN) represent two deneddylases which remove the ubiquitin-like Nedd8 from modified target proteins and are required for distinct fungal developmental programmes. The cellular DenA/DEN1 population is divided into a nuclear and a cytoplasmatic subpopulation which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service