Skip to Content
Merck
All Photos(1)

Key Documents

03-0190-00

Millipore

SMC® Human IL-18 High Sensitivity Immunoassay Kit

Synonym(s):

IGIF immunoassay kit, IL-18 Single Molecule Counting Kit, IL-18 ultra-sensitive immunoassay, Interleukin-18 immunoassay kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

species reactivity

mouse, feline, rat

manufacturer/tradename

SMC®

assay range

sensitivity: 0.09 pg/mL
(Lower Limit of Quantification)

inter-assay cv: 7%
intra-assay cv: 4%
(five normal plasma samples run in triplicate by 3 different operators on 3 different days.)

technique(s)

single molecule counting: suitable

storage temp.

2-8°C

General description

The SMC® Human IL-18 High Sensitivity Immunoassay Kit contains all reagents required to perform the assay. This assay takes advantage of a standard immunoassay workflow configured in a standard plate format.
Assay protocols are similar to existing sandwich ELISA methods with two key differences:
1) Elution buffer disrupts the sandwich, separating the labeled detection antibody for quantification.
2) The Erenna and SMCxPRO® immunoassay systems detect analytes using Single Molecule Counting (SMC®) technology.

Specificity

Species Reactivity:
Specificity to the following species recombinant proteins were tested with the following results:
Feline – 0.5% Mouse – 4% Rat – 0.3%

Cross Reactivity:
No cross-reactivity was seen with the following cytokines: IL-1a, IL-1ß, IL-1RA, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-10, IL-12 (p40), IL-12 (p70), IL-13, IL-15, IL-17A, IL-17E, IL-7F, IL-22, IL-27.

Accuracy:
  • Minimum of 5 serum samples spiked with average recovery of 99% with a range of 92-107%
  • Minimum of 5 plasma samples spiked with average recovery of 95% with a range of 88-103%

Application

The sensitivity of the Single Molecule Counting (SMC®) technology gives the SMC® Human IL-18 High Sensitivity Immunoassay Kit unparalleled performance in the quantification of IL-18 in human serum and plasma.

This new streamline kit has the following features including:
  • Improved options for sample prep and incubation
  • Lyophilization of the standard provides the kit at one storage condition
  • Increased reagent volumes to accommodate automation
  • Validated to run on both SMCxPRO® and Erenna


Research Category:
Inflammation & Immunology

Research Sub-Category:
  • Inflammatory Disease,
  • Immune Response
  • Autoimmune disease

Packaging

96 well assay

Components

  • Assay Buffer
  • IL-18 Coated Beads
  • Standard Diluent
  • IL-18 Detection Antibody
  • IL-18 Standard
  • 10X Wash Buffer
  • Buffer D
  • Elution Buffer B

Legal Information

SMC is a registered trademark of Singulex
SMCxPRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Use the power of high sensitivity immunology assays with SMC® technology to precisely characterize low-abundant cytokines like those from Th1/Th2 and Th17 cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service