MAN372903108
Manchester Organics
3-Bromo-5,6,7,8-tetrahydroimidazo[1,2-a]pyridine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
3-bromo-5,6,7,8-tetrahydroimidazo[1,2-a]pyridine
Inchi Code
1S/C7H9BrN2/c8-6-5-9-7-3-1-2-4-10(6)7/h5H,1-4H2
InChI key
CDQPJEFLGBLJKS-UHFFFAOYSA-N
Purity
95%
Country of Origin
GB
Product Link
https://www.manchesterorganics.com/B23515
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service