Skip to Content
Merck
All Photos(1)

Key Documents

KEY465195204

Key Organics / BIONET

4-(butan-2-yl)-2-[(E)-(hydroxyimino)methyl]phenol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
193.25
MDL number:

Pricing and availability is not currently available.

Iupac Name

5-sec-butyl-2-hydroxybenzaldehyde oxime

Inchi Code

1S/C11H15NO2/c1-3-8(2)9-4-5-11(13)10(6-9)7-12-14/h4-6,8,13H,3,7H2,1-2H3

InChI key

LTYCIXKZBLDAHH-UHFFFAOYSA-N

Purity

90%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/3R-0104

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

MSDS

https://www.keyorganics.net/sds/3R-0104


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service