Skip to Content
Merck
All Photos(1)

Key Documents

KEY375227039

Key Organics / BIONET

methyl 6-chloro-1-benzofuran-2-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
210.62
MDL number:

mp

97 - 99

Iupac Name

methyl 6-chloro-1-benzofuran-2-carboxylate

Inchi Code

1S/C10H7ClO3/c1-13-10(12)9-4-6-2-3-7(11)5-8(6)14-9/h2-5H,1H3

InChI key

BBKNMQLOVMQZRO-UHFFFAOYSA-N

Purity

97%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/LG-0740

Melting Point

97 - 99

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

MSDS

https://www.keyorganics.net/sds/LG-0740


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service