Skip to Content
Merck
All Photos(1)

Key Documents

KEY370561087

Key Organics / BIONET

5-bromo-2-phenylpyridine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
234.1
MDL number:
Pricing and availability is not currently available. Contact Technical Service

bp

309.3C at 760 mmHg

mp

73-75

Iupac Name

5-bromo-2-phenylpyridine

Inchi Code

1S/C11H8BrN/c12-10-6-7-11(13-8-10)9-4-2-1-3-5-9/h1-8H

InChI key

PRNGIODVYLTUKH-UHFFFAOYSA-N

storage temp.

Store under inert gas

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/BS-2050

Boiling point

309.3C at 760 mmHg

Melting Point

73-75

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service