Skip to Content
Merck

KEY189464737

Key Organics / BIONET

3-(tert-butoxy)benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
194.23
MDL number:

Pricing and availability is not currently available.

Iupac Name

3-tert-butoxybenzoic acid

Inchi Code

1S/C11H14O3/c1-11(2,3)14-9-6-4-5-8(7-9)10(12)13/h4-7H,1-3H3,(H,12,13)

InChI key

IURSJORDCNZJFE-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
HPA031369HPA043359HPA038600
biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

Gene Information

human ... CPNE3(8895)

Gene Information

human ... CPNE5(57699)

Gene Information

human ... CPNE7(27132)

Gene Information

human ... CPNE8(144402)

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/12P-016

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

MSDS

https://www.keyorganics.net/sds/12P-016


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service