JWPH3249E63F
J & W Pharmlab LLC
3,5-Dichloro-4-methoxy-pyridazine
About This Item
Iupac Name
3,5-dichloro-4-methoxypyridazine
Inchi Code
1S/C5H4Cl2N2O/c1-10-4-3(6)2-8-9-5(4)7/h2H,1H3
InChI key
FZBKNXQRYVZIKH-UHFFFAOYSA-N
storage temp.
Purity
Country of Origin
Physical Form
Product Link
MSDS
https://www.jwpharmlab.com/home/product/70R0260
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service