JWPH3249D04F
J & W Pharmlab LLC
3-tert-Butoxycarbonylmethyl-1H-indazole-6-carboxylic acid methyl ester
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
methyl 3-(2-(tert-butoxy)-2-oxoethyl)-1H-indazole-6-carboxylate
Inchi Code
1S/C15H18N2O4/c1-15(2,3)21-13(18)8-12-10-6-5-9(14(19)20-4)7-11(10)16-17-12/h5-7H,8H2,1-4H3,(H,16,17)
InChI key
PGGZIIZMNLMYPG-UHFFFAOYSA-N
Purity
96%
Country of Origin
US
Physical Form
White Solid
Product Link
https://www.jwpharmlab.com/home/product/45R0673
MSDS
https://www.jwpharmlab.com/home/product/45R0673
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service