Skip to Content
Merck
All Photos(1)

Key Documents

ENAH0385D47A

Enamine

tert-butyl (2R)-2-(2-methoxyacetyl)pyrrolidine-1-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$5.75
25 G
$9.20
100 G
$11.50
500 G
$25.30
1 KG
$46.00

Vendor SKU: A145351-5G

$5.75


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
5 G
$5.75
25 G
$9.20
100 G
$11.50
500 G
$25.30
1 KG
$46.00

About This Item

CAS Number:
Molecular Weight:
243.3
MDL number:

Vendor SKU: A145351-5G

$5.75


Ships in 7 days - from Aldrich Partner.

Iupac Name

tert-butyl (R)-2-(2-methoxyacetyl)pyrrolidine-1-carboxylate

Inchi Code

1S/C12H21NO4/c1-12(2,3)17-11(15)13-7-5-6-9(13)10(14)8-16-4/h9H,5-8H2,1-4H3/t9-/m1/s1

InChI key

YDTHZSZTWFGIBX-SECBINFHSA-N

storage temp.

4

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-799076

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-799076


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service