ENA333558728
Enamine
4-chloro-6-methoxyquinoline-3-carbonitrile
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
mp
226-227
Iupac Name
4-chloro-6-methoxy-3-quinolinecarbonitrile
Inchi Code
1S/C11H7ClN2O/c1-15-8-2-3-10-9(4-8)11(12)7(5-13)6-14-10/h2-4,6H,1H3
InChI key
VIKNILFXAQPQEF-UHFFFAOYSA-N
storage temp.
-10
Purity
95
Country of Origin
UA
Shipping Temperature
Ice Pack
Physical Form
powder
Product Link
https://www.enaminestore.com/catalog/EN300-83048
Melting Point
226-227
Signal Word
Warning
Hazard Statements
MSDS
https://www.enaminestore.com/catalog/EN300-83048
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service