Skip to Content
Merck

ENA161209133

Enamine

3-[4-(2-chloroacetyl)piperazin-1-yl]-1lambda6-thiolane-1,1-dione hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
317.24
MDL number:

Pricing and availability is not currently available.

Iupac Name

1-(chloroacetyl)-4-(1,1-dioxidotetrahydro-3-thienyl)piperazine hydrochloride

Inchi Code

1S/C10H17ClN2O3S.ClH/c11-7-10(14)13-4-2-12(3-5-13)9-1-6-17(15,16)8-9;/h9H,1-8H2;1H

InChI key

CZQDFRIVQWVIQK-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
EHU127751EHU018661EHU002301
Gene Information

human ... IMPDH2(3615), IMPDH2(3615)

Gene Information

human ... IMPDH1(3614), IMPDH1(3614)

Gene Information

human ... IMPG1(3617), IMPG1(3617)

Gene Information

human ... ITGAV(3685), ITGAV(3685)

product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

esiRNA cDNA target sequence

GCAGCAAGAAGGGAAAGTTGCCCATTGTAAATGAAGATGATGAGCTTGTGGCCATCATTGCCCGGACAGACCTGAAGAAGAATCGGGACTACCCACTAGCCTCCAAAGATGCCAAGAAACAGCTGCTGTGTGGGGCAGCCATTGGCACTCATGAGGATGACAAGTATAGGCTGGACTTGCTCGCCCAGGCTGGTGTGGATGTAGTGGTTTTGGACTCTTCCCAGGGAAATTCCATCTTCCAGATCAATATGATCAAGTACATCAAAGACAAATACCCTAATCTCCAAGTCATTGGAGGCAATGTGGTCACTGCTGCCCAGGCCAAGAACCTCATTGATGCAGGTGTGGATGCCCTGCGGGTGGGCATGGGAAGTGGCTCCATCTGCATTACGCAGGAAGTGCTGGCCTGTGGGCGGCCCCAAGCAACAGCAGTGTACAAGGTGTCAGAGTATGCACGGCGCTTTGGTGTTCCGGTCATTGCTGAT

esiRNA cDNA target sequence

ACCTCCCGAGACATCGACTTTCTTGCTGAGAAGGACCACACCACCCTCCTCAGTGAGGTGATGACGCCAAGGATTGAACTGGTGGTGGCTCCAGCAGGTGTGACGTTGAAAGAGGCAAATGAGATCCTGCAGCGTAGCAAGAAAGGGAAGCTGCCTATCGTCAATGATTGCGATGAGCTGGTGGCCATCATCGCCCGCACCGACCTGAAGAAGAACCGAGACTACCCTCTGGCCTCCAAGGATTCCCAGAAGCAGCTGCTCTGTGGGGCAGCTGTGGGCACCCGTGAGGATGACAAATACCGTCTGGACCTGCTCACCCAGGCGGGCGTCGACGTCATAGTCTTGGACTCGTCCCAAGGGAATTCGGTGTATCAGATCGCCATGGTGCATTACATCAAACAGAAGTACCCCCACCTCC

esiRNA cDNA target sequence

CTCCCTGTCAGAAGCTCCACCTTTCTTTATGGCATCAAGCATCTTCTCTCTGACTGATCAAGGCACCACAGATACAATGGCCACTGACCAGACAATGCTAGTACCAGGGCTCACCATCCCCACCAGTGATTATTCTGCAATCAGCCAACTGGCTCTGGGAATTTCACATCCACCTGCATCTTCAGATGACAGCCGATCAAGTGCAGGTGGCGAAGATATGGTCAGACACCTAGATGAAATGGATCTGTCTGACACTCCTGCCCCATCTGAGGTACCAGAGCTCAGCGAATATGTTTCTGTCCCAGATCATTTCTTGGAGGATACCACTCCTGTCTCAGCTTTACAGTATATCACCACTAGTTCTATGACCATTGCCCCCAAGGGCCGAGAGCTGGTAGTGTTCTTCAGTCTGCGTGTTGCTAACATGGCCTTCTCCAACGACCTGTTCAACAAGAGCTCTCTGGAGTACCGAGCTCTGGAGCAACAATTCACACAGCTGCTGG

esiRNA cDNA target sequence

ATTCCACTGCAGGCTGATTTCATCGGGGTTGTCCGAAACAATGAAGCCTTAGCAAGACTTTCCTGTGCATTTAAGACAGAAAACCAAACTCGCCAGGTGGTATGTGACCTTGGAAACCCAATGAAGGCTGGAACTCAACTCTTAGCTGGTCTTCGTTTCAGTGTGCACCAGCAGTCAGAGATGGATACTTCTGTGAAATTTGACTTACAAATCCAAAGCTCAAATCTATTTGACAAAGTAAGCCCAGTTGTATCTCACAAAGTTGATCTTGCTGTTTTAGCTGCAGTTGAGATAAGAGGAGTCTCGAGTCCTGATCATGTCTTTCTTCCGATTCCAAACTGGGAGCACAAGGAGAACCCTGAGACTGAAGAAGATGTTGGGCCAGTTGTTCAGCACATCTATGAGCTGAGAAACAATGGTCCAAGTTCATTCAGCAAGGCA

NCBI accession no.

NM_000884

NCBI accession no.

NM_000883

NCBI accession no.

NM_001563

NCBI accession no.

NM_002210

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

storage temp.

RT

Salt Data

Cl

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-22641

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-22641


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service