Skip to Content
Merck
All Photos(1)

Key Documents

COM448685137

Combi-Blocks, Inc.

3-Cyano-5-(4-cyanophenyl)phenol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
220.23
MDL number:

Pricing and availability is not currently available. Contact Technical Service

Iupac Name

5-hydroxy[1,1'-biphenyl]-3,4'-dicarbonitrile

Inchi Code

1S/C14H8N2O/c15-8-10-1-3-12(4-2-10)13-5-11(9-16)6-14(17)7-13/h1-7,17H

InChI key

DIJDJBUVDVMXFR-UHFFFAOYSA-N

Purity

96%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?YA-3091

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?YA-3091


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service