Skip to Content
Merck
All Photos(1)

Key Documents

C12H316CF138

Chemcia Scientific, LLC

4-Chloro-5-iodo-2-methyl-benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
296.49
MDL number:
Pricing and availability is not currently available. Contact Technical Service

Iupac Name

4-chloro-5-iodo-2-methylbenzoic acid

Inchi Code

1S/C8H6ClIO2/c1-4-2-6(9)7(10)3-5(4)8(11)12/h2-3H,1H3,(H,11,12)

InChI key

JFISQRPYNOQTAD-UHFFFAOYSA-N

storage temp.

0-5C

Purity

97%

Country of Origin

US

Shipping Temperature

Ambient

Physical Form

White Solid

Product Link

http://www.chemcia.com/product_fl02.asp?id=6755

MSDS

http://www.chemcia.com/product_fl02.asp?id=6755


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service