Skip to Content
Merck
All Photos(1)

Key Documents

CH4017936920

ChemBridge Corporation

[(4,6-diaminopyrimidin-2-yl)thio]acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
200.22
MDL number:
Pricing and availability is not currently available. Contact Technical Service

Iupac Name

[(4,6-diamino-2-pyrimidinyl)sulfanyl]acetic acid

Inchi Code

1S/C6H8N4O2S/c7-3-1-4(8)10-6(9-3)13-2-5(11)12/h1H,2H2,(H,11,12)(H4,7,8,9,10)

InChI key

JKAISOKGXPAILZ-UHFFFAOYSA-N

storage temp.

RT

Purity

95%

Country of Origin

KZ

Shipping Temperature

RT

Physical Form

solid

Product Link

https://www.hit2lead.com/building-blocks/6549621

MSDS

https://www.hit2lead.com/sds/6549621_sds.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service