Skip to Content
Merck
All Photos(1)

Documents

C-261

Supelco

Crotonaldehyde-2,4-DNPH

certified reference material, vial of 100 mg, Cerilliant®

Synonym(s):

Crotonaldehyde-2,4-dinitrophenylhydrazone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCH=NNHC6H3(NO2)2
CAS Number:
Molecular Weight:
250.21
MDL number:
UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

vial of 100 mg

manufacturer/tradename

Cerilliant®

mp

192-195 °C (lit.)

format

neat

storage temp.

-10 to -25°C

SMILES string

C\C=C\C=N\Nc1ccc(cc1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C10H10N4O4/c1-2-3-6-11-12-9-5-4-8(13(15)16)7-10(9)14(17)18/h2-7,12H,1H3/b3-2+,11-6+

InChI key

GFUVNGJSSAEZHW-PDGXAJGZSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service