Skip to Content
Merck
All Photos(2)

Documents

850850P

Avanti

18:1 Monomethyl PE

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-methyl, powder

Synonym(s):

1,2-di-(9Z-octadecenoyl)-sn-glycero-3-phosphoethanolamine-N-methyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H80NO8P
CAS Number:
Molecular Weight:
758.06
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (850850P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850850P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[N+](C)([H])[H])=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O

InChI

1S/C42H80NO8P/c1-4-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-41(44)48-38-40(39-50-52(46,47)49-37-36-43-3)51-42(45)35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-5-2/h18-21,40,43H,4-17,22-39H2,1-3H3,(H,46,47)/b20-18+,21-19+/t40-/m1/s1

InChI key

LPXFOQGBESUDAX-KFVQOHGZSA-N

General description

18:1 Monomethyl phosphoethanolamine (PE) is an alkyl phosphate derivative. Methyl transferases catalyzes the formation of monomethyl PE by acting on PE. It might be involved in the transduction of biochemical signals via the cell membrane.

Application

18:1 Monomethyl PE may be used for the preparation of standard liposomes. It is also suitable to study its effect on the interaction of fructan with monomolecular lipid system.

Packaging

5 mL Clear Glass Sealed Ampule (850850P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Membrane Fluidity (2012)
Structural requirements of the fructan-lipid interaction
Vereyken IJ, et al.
Biophysical Journal, 84(5), 3147-3154 (2003)
Effects of lipids on the interaction of SecA with model membranes
Ahn T, et al.
Archives of Biochemistry and Biophysics, 395(1), 14-20 (2001)
Grzegorz Pawlik et al.
The Journal of biological chemistry, 295(8), 2473-2482 (2020-01-15)
Phospholipid N-methyltransferases (PLMTs) synthesize phosphatidylcholine by methylating phosphatidylethanolamine using S-adenosylmethionine as a methyl donor. Eukaryotic PLMTs are integral membrane enzymes located in the endoplasmic reticulum (ER). Recently Opi3, a PLMT of the yeast Saccharomyces cerevisiae was proposed to perform in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service