ATC429238442
Atlantic Research Chemicals Ltd
5-Nitrothiophene-2-carbonyl chloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
5-nitro-2-thiophenecarbonyl chloride
Inchi Code
1S/C5H2ClNO3S/c6-5(8)3-1-2-4(11-3)7(9)10/h1-2H
InChI key
BYEANWQDRDXPPA-UHFFFAOYSA-N
Purity
95
Country of Origin
GB
Product Link
https://atlantic-chemicals.com/product/?code=CC00316
MSDS
https://atlantic-chemicals.com/product/?code=CC00316
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service