ATE333216084
AstaTech, Inc.
(R)-2-(PIPERIDIN-3-YL)ACETIC ACID HCL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
(3R)-3-piperidinylacetic acid hydrochloride
Inchi Code
1S/C7H13NO2.ClH/c9-7(10)4-6-2-1-3-8-5-6;/h6,8H,1-5H2,(H,9,10);1H/t6-;/m1./s1
InChI key
IMUCGEDHTPKPBM-FYZOBXCZSA-N
storage temp.
2-8 C
Purity
97%
Shipping Temperature
Normal
Physical Form
Pale-yellow to Yellow-brown Solid
Product Link
http://astatechinc.com/CPSResult.php?CRNO=4990
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
http://astatechinc.com/Asta_MSDS.php?Catalog=56293
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service