APO455834986
Apollo Scientific Ltd
2-Chloro-3,4,4,4-tetrafluoro-3-(trifluoromethyl)but-1-ene
Sign Into View Organizational & Contract Pricing
Select a Size
All Photos(1)
Select a Size
Change View
About This Item
Iupac Name
2-chloro-3,4,4,4-tetrafluoro-3-(trifluoromethyl)-1-butene
Inchi Code
1S/C5H2ClF7/c1-2(6)3(7,4(8,9)10)5(11,12)13/h1H2
InChI key
VJFYSORKRLINOQ-UHFFFAOYSA-N
storage temp.
Ambient Temperature
Purity
97%
Country of Origin
GB
Shipping Temperature
Ambient Temperature
Physical Form
Liquid
Product Link
https://store.apolloscientific.co.uk/product/2-chloro-3444-tetrafluoro-3-trifluoromethylbut-1-ene
Synonym
1H,1H-Heptafluoro(2-chloro-3-methylbut-1-ene)
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://store.apolloscientific.co.uk/product/2-chloro-3444-tetrafluoro-3-trifluoromethylbut-1-ene
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service