Skip to Content
Merck
All Photos(1)

Key Documents

AOBH30376740

AOBChem

(2-Fluoro-6-methoxy-4-methylphenyl)methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
170.18
MDL number:
Pricing and availability is not currently available. Contact Technical Service

Iupac Name

(2-fluoro-6-methoxy-4-methylphenyl)methanol

Inchi Code

1S/C9H11FO2/c1-6-3-8(10)7(5-11)9(4-6)12-2/h3-4,11H,5H2,1-2H3

InChI key

AHVAHMZOILSUFA-UHFFFAOYSA-N

storage temp.

2-8C

Purity

97

Country of Origin

CN

Shipping Temperature

Room Temperature

Physical Form

Solid

Product Link

https://aobchem.com/2-fluoro-6-methoxy-4-methylphenyl-methanol.html

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://aobchem.com/pub/media/files/sds/34903.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service