Skip to Content
Merck
All Photos(1)

Documents

AMBH97BA1142

Ambeed, Inc.

2,5-Bis(2-octyldodecyl)-3,6-di(thiophen-2-yl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
861.44
MDL number:

Iupac Name

2,5-bis(2-octyldodecyl)-3,6-di(thiophen-2-yl)-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione

Inchi Code

1S/C54H88N2O2S2/c1-5-9-13-17-21-23-27-31-37-45(35-29-25-19-15-11-7-3)43-55-51(47-39-33-41-59-47)49-50(53(55)57)52(48-40-34-42-60-48)56(54(49)58)44-46(36-30-26-20-16-12-8-4)38-32-28-24-22-18-14-10-6-2/h33-34,39-42,45-46H,5-32,35-38,43-44H2,1-4H3

InChI key

LTFXMPXYDRPSSF-UHFFFAOYSA-N

storage temp.

Keep in dark place,Sealed in dry,Room Temperature

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/1267540-02-2.html

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.ambeed.com/products/1267540-02-2.html


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service