Skip to Content
Merck
All Photos(1)

Key Documents

AMBH9614F89E

Ambeed, Inc.

1-Boc-4-Ethylaminopiperidine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
228.33
MDL number:

Iupac Name

tert-butyl 4-(ethylamino)-1-piperidinecarboxylate

Inchi Code

1S/C12H24N2O2/c1-5-13-10-6-8-14(9-7-10)11(15)16-12(2,3)4/h10,13H,5-9H2,1-4H3

InChI key

SRDSJYNZSNVSGS-UHFFFAOYSA-N

storage temp.

Keep in dark place,Inert atmosphere,Room temperature

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Solid or semi-solid or lump or liquid

Product Link

https://www.ambeed.com/products/264905-39-7.html

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Precautionary statements


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service