Key Documents
Z687235
Sigma-Aldrich
Primary vapor removal filter module with pleated after-filter for Aldrich® compact ductless air cleaning system
activated alumina with potassium permanganate
About This Item
UNSPSC Code:
41100000
NACRES:
NB.43
description
refillable
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)