WHA800309
Whatman® polycarbonate Nuclepore filter discs 0.1 μm pore size
hydrophilic, track-etched, treated with PVP, 100 ea, 19 mm diam
Synonym(s):
Z747130, nuclepore membrane, polycarbonate membrane
Select a Size
Select a Size
About This Item
Recommended Products
Product Name
Whatman® Nuclepore™ Track-Etched Membranes, diam. 19 mm, pore size 0.1 μm, polycarbonate
material
polycarbonate
sterility
non-sterile
packaging
pkg of 100 ea
manufacturer/tradename
Whatman 800309
Whatman Article No. 28421046 (US reference)
diam.
19 mm
pore size
0.1 μm
Looking for similar products? Visit Product Comparison Guide
Application
- Epifluorescence microscopy
- Environmental analysis
- Cell biologyEPA testing
- Fuel testing
- Bioassays
- Parasitology
- Air analysis
- Water microbiology
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service