Skip to Content
Merck
All Photos(1)

Documents

W311901

Sigma-Aldrich

Ylang-ylang oil

FG

Synonym(s):

Ylang-ylang oil III

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
3119
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21

biological source

Cananga odorata
synthetic

Quality Level

grade

FG
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012

optical activity

[α]20/D −38°, neat

refractive index

n20/D 1.508

bp

261 °C/760 mmHg

density

0.928 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

floral

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Taste at 3 ppm

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 27, 92-92 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service