Skip to Content
Merck
All Photos(1)

Documents

T66001

Sigma-Aldrich

Naringenin

Synonym(s):

(±)-Naringenin, (±)-2,3-Dihydro-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 4′,5,7-Trihydroxyflavanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12O5
CAS Number:
Molecular Weight:
272.25
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

mp

247-250 °C (lit.)

SMILES string

OC1=CC=C(C=C1)C(C2)OC3=CC(O)=CC(O)=C3C2=O

InChI

1S/C15H12O5/c16-9-3-1-8(2-4-9)13-7-12(19)15-11(18)5-10(17)6-14(15)20-13/h1-6,13,16-18H,7H2

InChI key

FTVWIRXFELQLPI-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nada Oršolić et al.
European journal of nutrition, 53(5), 1217-1227 (2013-11-26)
Reactive oxygen species play a role in a number of degenerative conditions including osteoporosis. Flavonoids as phyto-oestrogens exert physiological effects against oxidative stress diseases. We developed a retinoic acid-induced bone loss model of rats to assess whether flavonoids and alendronate
Senem Kamiloglu et al.
Journal of the science of food and agriculture, 94(11), 2225-2233 (2014-01-01)
In order to investigate the effect of home processing on the bioaccessibility of health-related constituents of tomatoes, total lycopene, phenolics, flavonoids and antioxidant capacity were determined from seven different tomato products using an in vitro gastrointestinal digestion model. Additionally, the
Julia M Assini et al.
Endocrinology, 156(6), 2087-2102 (2015-03-17)
The molecular mechanisms and metabolic pathways whereby the citrus flavonoid, naringenin, reduces dyslipidemia and improves glucose tolerance were investigated in C57BL6/J wild-type mice and fibroblast growth factor 21 (FGF21) null (Fgf21(-/-)) mice. FGF21 regulates energy homeostasis and the metabolic adaptation
Manuel Y Schär et al.
The American journal of clinical nutrition, 101(5), 931-938 (2015-03-20)
Epidemiologic data suggest inverse associations between citrus flavanone intake and cardiovascular disease (CVD) risk. However, insufficient randomized controlled trial data limit our understanding of the mechanisms by which flavanones and their metabolites potentially reduce cardiovascular risk factors. We examined the
S Bhattacharya et al.
Diabetes, obesity & metabolism, 16(7), 602-612 (2013-11-12)
Caffeic acid, naringenin and quercetin are naturally occurring phenolic compounds (PCs) present in many plants as secondary metabolites. The aim of this study was to investigate their effect on glucose-stimulated insulin secretion (GSIS) in INS-1E cells and to explore their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service