Skip to Content
Merck
All Photos(1)

Documents

SYNC362630

Synthware chromatography column with PTFE stopcock

I.D. 20.0 mm, L 305 mm

Synonym(s):

Synthware C362630, glass chromatograpy column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.51

grade

for chromatography

packaging

pkg of 1 ea

manufacturer/tradename

Synthware C362630

I.D.

20.0 mm

L

305 mm

Looking for similar products? Visit Product Comparison Guide

General description

With reinforced beaded top. All Synthware columns are heavy wall designed to increase safety.

Legal Information

Synthware is a trademark of Kemtech America Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service