Skip to Content
Merck
All Photos(2)

Key Documents

M54104

Sigma-Aldrich

Methyl DL-mandelate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OH)COOCH3
CAS Number:
Molecular Weight:
166.17
Beilstein:
2047558
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

bp

135 °C/12 mmHg (lit.)

mp

54-56 °C (lit.)

SMILES string

COC(=O)C(O)c1ccccc1

InChI

1S/C9H10O3/c1-12-9(11)8(10)7-5-3-2-4-6-7/h2-6,8,10H,1H3

InChI key

ITATYELQCJRCCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jörgen Samuelsson et al.
Journal of chromatography. A, 1203(2), 177-184 (2008-08-02)
The calculation of the adsorption energy distribution (AED) was recently introduced as an important tool for the chromatographic community for characterization of modern phases. The AED-calculations, provides model-independent information about the numbers of different adsorption sites and their respective energy-levels
Han-Ning Wei et al.
Applied biochemistry and biotechnology, 149(1), 79-88 (2008-03-20)
Microorganisms producing lipase were isolated from soil and sewage samples and screened for enantioselective resolution of (R,S)-methyl mandelate to (R)-mandelic acid. A strain designated as GXU56 was obtained and identified as Burkholderia sp. Preparing immobilized GXU56 lipase by simple adsorption
Y Chiang et al.
The Journal of organic chemistry, 65(4), 1175-1180 (2000-05-18)
Flash photolysis of methyl phenyldiazoacetate in aqueous solution produced phenylcarbomethoxycarbene, whose hydration generated a short-lived transient species that was identified as the enol isomer of methyl mandelate. This assignment is supported by the shape of the rate profile for decay
Fei Guo et al.
Applied microbiology and biotechnology, 97(8), 3355-3362 (2012-11-28)
Despite directed evolution being a practical and efficient method of improving the properties of enzymes, a trade-off between the targeted property and other essential properties often exists which hinders the efficiency of directed evolution. In our previous work, mutant CVH
Jie-Hua Shi et al.
Journal of molecular modeling, 18(2), 803-813 (2011-05-20)
Host-guest interactions of permethylated β-cyclodextrin (PM-β-CD) with methyl mandelate enantiomers ((R/S)-MMA) were simulated using semiempirical PM3 and ONIOM (B3LYP/6-31G(d):PM3) method. The chiral recognition mechanism of (R/S)-MMA enantiomers on PM-β-CD was investigated. The binding energies for all orientations considered in this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service