Skip to Content
Merck
All Photos(2)

Key Documents

GF87514421

Tantalum

rod, 50mm, diameter 12.7mm, annealed, 99.9%

Synonym(s):

Tantalum, TA007942, Ta

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ta
CAS Number:
Molecular Weight:
180.95
MDL number:
UNSPSC Code:
12141741
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.01 mmHg ( 537.2 °C)

Assay

≥99.9%

form

rod

autoignition temp.

572 °F

manufacturer/tradename

Goodfellow 875-144-21

resistivity

13.5 μΩ-cm, 20°C

bp

5425 °C (lit.)

mp

2996 °C (lit.)

density

16.69 g/cm3 (lit.)

SMILES string

[Ta]

InChI

1S/Ta

InChI key

GUVRBAGPIYLISA-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-kang Tang et al.
Zhongguo gu shang = China journal of orthopaedics and traumatology, 26(7), 617-620 (2013-10-19)
Incorrect treatment for femur head necrosis can cause collapse of femoral head and tresult in severe harm for the patients (especially for the patient with middle-aged and young). The structure and mechanics characteristics of tantalum rod is similar to bone
Ulrich Dorn et al.
The Journal of arthroplasty, 29(4), 831-835 (2013-10-09)
This study compared the corrosion behavior of tantalum-coated cobalt-chromium modular necks with that of titanium alloy modular necks at their junction to titanium-alloy femoral stem. Tests were performed in a dry assembly and two wet assemblies, one contaminated with calf
[Tantalum dust as a contrast substance for laryngography and bronchography (a review of the literature)].
R M Rabinovich
Vestnik rentgenologii i radiologii, (1)(1), 77-79 (1981-01-01)
Clinical use of porous tantalum in complex primary total knee arthroplasty.
Neil P Sheth et al.
American journal of orthopedics (Belle Mead, N.J.), 38(10), 526-530 (2009-12-17)
Brett Levine et al.
The journal of knee surgery, 20(3), 185-194 (2007-08-02)
Porous tantalum represents an alternative metal for primary and revision total knee arthroplasty (TKA) with several unique properties. Tantalum is a transition metal, which in its bulk form has shown excellent biocompatibility and is safe to use in vivo as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service