Skip to Content
Merck
All Photos(2)

Key Documents

GF59305237

Silver

insulated wire, 2m, conductor diameter 0.075mm, insulation thickness 0.010mm, nylon insulation, 99.99%

Synonym(s):

Silver, AG005811

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag
CAS Number:
Molecular Weight:
107.87
MDL number:
UNSPSC Code:
12141740
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99%

form

wire

manufacturer/tradename

Goodfellow 593-052-37

resistivity

1.59 μΩ-cm, 20°C

L × diam. × thickness

2 m × 0.075 mm × 0.010 mm

bp

2212 °C (lit.)

mp

960 °C (lit.)

density

10.49 g/cm3 (lit.)

SMILES string

[Ag]

InChI

1S/Ag

InChI key

BQCADISMDOOEFD-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neelam Gogoi et al.
Journal of nanoscience and nanotechnology, 14(6), 4147-4155 (2014-04-18)
In this work storage of silver nanoparticles (Ag NPs) in chitosan gel and its subsequent release for catalytic reduction processes is investigated. The generation of small sized metal nanoparticles which acts as catalyst is prerequisite to progress of a catalytic
Paulraj Kanmani et al.
Food chemistry, 148, 162-169 (2013-11-23)
Active nanocomposite films were prepared by blending aqueous solutions of gelatin with different concentrations of silver nanoparticles (AgNPs) using a solvent casting method. Formation of silver nanoparticles in the solution and films was confirmed with the surface plasmon resonance (SPR)
Takayoshi Kakuta et al.
Journal of nanoscience and nanotechnology, 14(6), 4090-4096 (2014-04-18)
Three-typed porphyrin derivatives with a different chain-length alkylcarboxylic acid as their peripheral anchor group have been prepared. Anodic photocurrents were observed in a simple system where the porphyrin derivatives were directly anchored on an indium tin oxide (ITO) electrode. Cathodic
S R Mueller-Spitz et al.
Letters in applied microbiology, 58(4), 330-337 (2013-11-30)
Polycyclic aromatic hydrocarbons (PAH) are a common environmental contaminant originating from both anthropogenic and natural sources. Mycobacterium species are highly adapted to utilizing a variety of PAH. Silver nanoparticles (AgNP) are an emerging contaminant that possess bactericidal properties, interferes with
Ana López-Serrano Oliver et al.
Environmental pollution (Barking, Essex : 1987), 189, 87-91 (2014-03-20)
Silver nanoparticles (AgNPs) are widely used in many applications and likely released into the aquatic environment. There is increasing evidence that Ag is efficiently delivered to aquatic organisms from AgNPs after aqueous and dietary exposures. Accumulation of AgNPs through the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service