Skip to Content
Merck
All Photos(1)

Key Documents

D198609

Sigma-Aldrich

2,5-Dinitrophenol

97%

Synonym(s):

γ-Dinitrophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)2C6H3OH
CAS Number:
Molecular Weight:
184.11
Beilstein:
1913411
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

contains

5-25% water

technique(s)

titration: suitable

mp

103-106 °C (lit.)

SMILES string

Oc1cc(ccc1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C6H4N2O5/c9-6-3-4(7(10)11)1-2-5(6)8(12)13/h1-3,9H

InChI key

UWEZBKLLMKVIPI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L V Bronov et al.
Antibiotiki, 26(1), 24-28 (1981-01-01)
Possible use of dinitrophenol reaction with amines in quantitative determination of amines and amino sugars in various antibiotics was investigated. It was found that the reaction with 2,5-dinitrophenol (2,5-DNP) in dimethylsulfoxide (DMSO) providing the highest batochromic shift in the electron
Surendra Vikram et al.
PloS one, 8(12), e84766-e84766 (2014-01-01)
Biodegradation of para-Nitrophenol (PNP) proceeds via two distinct pathways, having 1,2,3-benzenetriol (BT) and hydroquinone (HQ) as their respective terminal aromatic intermediates. Genes involved in these pathways have already been studied in different PNP degrading bacteria. Burkholderia sp. strain SJ98 degrades

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service