Skip to Content
Merck
All Photos(3)

Key Documents

I29204

Sigma-Aldrich

Itaconic acid

≥99%

Synonym(s):

Methylenesuccinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2C(=CH2)CO2H
CAS Number:
Molecular Weight:
130.10
Beilstein:
1759501
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99%

autoignition temp.

1472 °F

mp

165-168 °C (lit.)

density

1.573 g/mL at 25 °C (lit.)

SMILES string

OC(=O)CC(=C)C(O)=O

InChI

1S/C5H6O4/c1-3(5(8)9)2-4(6)7/h1-2H2,(H,6,7)(H,8,9)

InChI key

LVHBHZANLOWSRM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Itaconic acid (IA) can be used:
  • As a comonomer in the polymerization of polyacrylonitrile (PAN) to promote the thermo-oxidative stabilization of polymer.
  • In combination with acrylamide to form (poly[acrylamide-co-(itaconicacid)]) to synthesize biodegradable superabsorbent polymers.
  • To synthesize biobased polyester composite in fabric industry.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechanism and kinetics of the stabilization reactions of itaconic acid-modified polyacrylonitrile
Ouyang Q, et al.
Polymer Degradation and Stability, 93(8), 1415-1421 (2008)
Graft copolymerization, characterization, and degradation of cassava starch-g-acrylamide itaconic acid superabsorbents
Lanthong P, et al.
Carbohydrate Polymers, 66(2), 229-245 (2006)
Selective and flexible transformation of biomass-derived platform chemicals by a multifunctional catalytic system.
Frank M A Geilen et al.
Angewandte Chemie (International ed. in English), 49(32), 5510-5514 (2010-06-30)
Reddithota J Krupadam et al.
Environmental science and pollution research international, 19(5), 1841-1851 (2011-12-31)
Microcystins (MCs; cyclic heptapeptides) are produced by freshwater cyanobacteria and cause public health concern in potable water supplies. There are more than 60 types of MCs identified to date, of which MC-LR is the most common found worldwide. For MC-LR
E Karadağ et al.
Biomaterials, 17(1), 67-70 (1996-01-01)
In this in vitro study, the biocompatibility of acrylamide-based hydrogels such as acrylamide/crotonic and acrylamide/itaconic acid, prepared by using gamma-rays, has been investigated against some biochemical parameters of human serum. The swelling kinetics of acrylamide/crotonic and acrylamide/itaconic acid hydrogels are

Articles

Our strategy is to synthesize mesoporous carbonaceous materials (“Starbons”) using mesoporous expanded starch as the precursor without the need for a templating agent.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service