Skip to Content
Merck
All Photos(1)

Documents

933414

Sigma-Aldrich

SEM Mounting

Aluminum Standard, 15X10mm, 10/PK

Synonym(s):

cylinder stub

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111761
NACRES:
NA.23

Looking for similar products? Visit Product Comparison Guide

General description

EMS techware

Application

Cylinder specimen mounts are for use with JEOL and ISI scanning electron microscopes.
Surface: ⅝" dia. (15mm), height ⅜" (9.5mm)
Aluminum: Standard
The mounts are made from ultra-pure aluminum with standard lathe finish.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service