Skip to Content
Merck
All Photos(1)

Key Documents

907731

Sigma-Aldrich

Benzoic acid, 4-isocyanato-ethylene glycol bis lactic acid ester

Synonym(s):

Absorbable isocyanate, Isocyanate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$85.20

$85.20


Estimated to ship onApril 19, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$85.20

About This Item

Empirical Formula (Hill Notation):
C24H20N2O10
Molecular Weight:
496.42
UNSPSC Code:
12352106
NACRES:
NA.23

$85.20


Estimated to ship onApril 19, 2025


Request a Bulk Order

form

powder or solid

color

white to yellow

storage temp.

−20°C

Application

Benzoic acid, 4-isocyanato-ethylene glycol bis lactic acid ester is a highly reactive absorbable isocyanate. The biodegradable linkage in this monomer is derived from biocompatible groups. This novel monomer enables the synthesis of biodegradable polyurethanes that possess comparable mechanical properties with commercially available medical grade polyurethanes.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service