Skip to Content
Merck
All Photos(1)

Documents

900179

Sigma-Aldrich

Monodispersed nanodiamond particles

5 nm avg. part. size (DLS), 10 mg/mL in DMSO, hydroxylated

Synonym(s):

Detonation Nanodiamond, Nanodiamond

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352302
NACRES:
NA.23

description

+34.5 mV ±20%

form

dispersion
nanoparticles

concentration

10 mg/mL in DMSO

avg. part. size

5 nm (DLS)

Looking for similar products? Visit Product Comparison Guide

Application

Detonation nanodiamonds find applications in polymer composites for providing improved mechanical properties, UV protection, and improved resistance to radiation. They also find biological applications extending from drug delivery to positron emission tomography and other imaging techniques.

Other Notes

Do not freeze the materials.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nanodiamond Particles: Properties and Perspectives for Bioapplications.
Schrand AM
Critical Reviews in Solid State and Materials Sciences, 34, 18-18 (2009)
The properties and applications of nanodiamonds.
Mochalin VN, et al.
Nature Nanotechnology, 7, 11-11 (2012)
Nanodiamond Particles: Properties and Perspectives for Bioapplications.
Schrand AM, et al.
Critical Reviews in Solid State and Materials Sciences, 34, 18-18 (2009)
The properties and applications of nanodiamonds.
Mochalin VN,et al.
Nature Nanotechnology, 7, 11-11 (2012)
Cancer nanomedicine: from drug delivery to imaging.
Chow E, et al.
Science Translational Medicine, 5 (2013)

Articles

Among various nanomaterials, nanoparticles of less than 10 nm size remain the most sought-after products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service