Skip to Content
Merck
All Photos(1)

Key Documents

775789

Sigma-Aldrich

8-Bromocaffeine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9BrN4O2
CAS Number:
Molecular Weight:
273.09
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

208-210 °C

SMILES string

CN1C(=O)N(C)c2nc(Br)n(C)c2C1=O

InChI

1S/C8H9BrN4O2/c1-11-4-5(10-7(11)9)12(2)8(15)13(3)6(4)14/h1-3H3

InChI key

YRLRORFORQUTKS-UHFFFAOYSA-N

Application

Promotes the conversion of aldoximes to nitriles under mildly basic conditions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D L Leonidze et al.
Radiobiologiia, 26(2), 231-235 (1986-03-01)
In experiments on mice with solid sarcoma 37, a study was made of modification of the radiosensitizing effect of metronidazole. The increase in the leucotic interferon titre, induced by the interferon synthesis inductors, did not influence the radiosensitizing efficiency of
[Radiation-modifying effect of 8-bromocaffeine on mouse La leukosis cells].
V A Volchkov et al.
Radiobiologiia, 22(2), 255-257 (1982-03-01)
[The radiation therapy of experimental brain tumors by using the radiosensitizing preparation xanthobin (8-bromocaffeine)].
L P Vartanian et al.
Meditsinskaia radiologiia, 34(11), 82-83 (1989-11-01)
[Effect of xanthobin on the degree of damage to DNA molecules by gamma-irradiation].
N D Osipov et al.
Radiobiologiia, 20(5), 671-675 (1980-09-01)
[Effect of 8-bromocaffeine on repair of sublethal injuries of murine hematopoietic stem cells after fractionated irradiation].
L P Vartanian et al.
Radiobiologiia, 22(5), 685-687 (1982-09-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service