Skip to Content
Merck
All Photos(1)

Key Documents

766488

Sigma-Aldrich

Methyl 3-bromo-4-iodobenzoate

97%

Synonym(s):

3-Bromo-4-iodobenzoic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6BrIO2
CAS Number:
Molecular Weight:
340.94
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

56-60 °C

storage temp.

2-8°C

SMILES string

COC(=O)c1ccc(I)c(Br)c1

InChI

1S/C8H6BrIO2/c1-12-8(11)5-2-3-7(10)6(9)4-5/h2-4H,1H3

InChI key

OIMBMPVAESWCEA-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service