Skip to Content
Merck
All Photos(1)

Key Documents

744417

Sigma-Aldrich

Phenyl cyanate solution

~20 wt. % in dichloromethane, ≥90% (GC)

Synonym(s):

Cyanatobenzene solution, Cyanic acid phenyl ester solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5NO
CAS Number:
Molecular Weight:
119.12
Beilstein:
507925
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥90% (GC)

form

liquid

concentration

~20 wt. % in dichloromethane

density

~1.27 g/mL at 20 °C

functional group

cyanate
phenoxy

storage temp.

−20°C

SMILES string

N#COc1ccccc1

InChI

1S/C7H5NO/c8-6-9-7-4-2-1-3-5-7/h1-5H

InChI key

CWHFDTWZHFRTAB-UHFFFAOYSA-N

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service