Skip to Content
Merck
All Photos(1)

Key Documents

725781

Sigma-Aldrich

Ethyl 3,3-difluorobutyrate

≥96.0%

Synonym(s):

3,3-Difluorobutyric acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10F2O2
CAS Number:
Molecular Weight:
152.14
Beilstein:
1759658
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96.0%

form

liquid

SMILES string

CCOC(=O)CC(C)(F)F

InChI

1S/C6H10F2O2/c1-3-10-5(9)4-6(2,7)8/h3-4H2,1-2H3

InChI key

JSMRUPPAFDYIRD-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

97.1 °F

Flash Point(C)

36.19 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service